MAN238930891 Manchester Organics

3-Chloro-5-nitrobenzaldehyde

  • MDL number MFCD08236780
  • Cas Number : 22233-54-1
  • Molecular Weight : 185.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-nitrobenzaldehyde
Inchi Code 1S/C7H4ClNO3/c8-6-1-5(4-10)2-7(3-6)9(11)12/h1-4H
Inchi Key KXGORBDLPVCCOJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J20611
Availability details for
Quantity
  • Availability
  • Add to Cart