MAN238930904 Manchester Organics

2-(Cyanophenyl)acetic acid

  • MDL number MFCD01646238
  • Molecular Weight : 161.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F20702

Safety Information

MSDS https://www.manchesterorganics.com/F20702
Availability details for
Quantity
  • Availability
  • Add to Cart