MAN238930913 Manchester Organics

4-Amino-2-fluorobenzoic acid tert-butyl ester

  • MDL number MFCD06658385
  • Molecular Weight : 211.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X21097

Safety Information

MSDS https://www.manchesterorganics.com/X21097
Availability details for
Quantity
  • Availability
  • Add to Cart