MAN238930971 Manchester Organics

1-Bromo-3-iodo-5-(trifluoromethoxy)benzene

  • MDL number MFCD06657955
  • Molecular Weight : 366.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N20876

Safety Information

MSDS https://www.manchesterorganics.com/N20876
Availability details for
Quantity
  • Availability
  • Add to Cart