MAN238930978 Manchester Organics

5-(2-Methoxy-phenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD05170022
  • Molecular Weight : 218.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A20768

Safety Information

MSDS https://www.manchesterorganics.com/A20768
Availability details for
Quantity
  • Availability
  • Add to Cart