MAN238930990 Manchester Organics

3-Bromo-2-phenyl-imidazo[1,2-a]pyrimidine

  • MDL number MFCD06660438
  • Cas Number : 904814-72-8
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B20769
Availability details for
Quantity
  • Availability
  • Add to Cart