MAN238931029 Manchester Organics

(2-Amino-2-phenyl-ethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD06245566
  • Cas Number : 174885-99-5
  • Molecular Weight : 236.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B20865
Availability details for
Quantity
  • Availability
  • Add to Cart