MAN238931030 Manchester Organics

1-Benzo[1,3]dioxol-5-yl-ethylamine

  • MDL number MFCD02656661
  • Molecular Weight : 165.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D20938

Safety Information

MSDS https://www.manchesterorganics.com/D20938
Availability details for
Quantity
  • Availability
  • Add to Cart