MAN238931047 Manchester Organics

2,4-Dichloro-1-[isocyano-(toluene-4-sulfonyl)-methyl]-benzene

  • MDL number MFCD04114807
  • Molecular Weight : 340.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z21383

Safety Information

MSDS https://www.manchesterorganics.com/Z21383
Availability details for
Quantity
  • Availability
  • Add to Cart