MAN238931049 Manchester Organics

3-(1H-Indol-2-yl)-phenylamine

  • MDL number MFCD00454976
  • Molecular Weight : 208.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E20985

Safety Information

MSDS https://www.manchesterorganics.com/E20985
Availability details for
Quantity
  • Availability
  • Add to Cart