MAN238931092 Manchester Organics

2-[(Propylamino)carbonyl]cyclohexanecarboxylic acid

  • MDL number MFCD03388374
  • Molecular Weight : 213.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E20394

Safety Information

MSDS https://www.manchesterorganics.com/E20394
Availability details for
Quantity
  • Availability
  • Add to Cart