MAN238931095 Manchester Organics

4-[(2-Isopropyl-6-methylphenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD06065527
  • Molecular Weight : 249.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G20396

Safety Information

MSDS https://www.manchesterorganics.com/G20396
Availability details for
Quantity
  • Availability
  • Add to Cart