MAN238931137 Manchester Organics

1-Phenyl-imidazolidin-2-one

  • MDL number MFCD02671066
  • Molecular Weight : 162.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H20942

Safety Information

MSDS https://www.manchesterorganics.com/H20942
Availability details for
Quantity
  • Availability
  • Add to Cart