MAN238931174 Manchester Organics

4-[(2-Fluoro-5-methylphenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD06059320
  • Molecular Weight : 225.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M20449

Safety Information

MSDS https://www.manchesterorganics.com/M20449
Availability details for
Quantity
  • Availability
  • Add to Cart