MAN238931196 Manchester Organics

1-Ethyl-1H-indole-2,3-dione

  • MDL number MFCD00224220
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R20498

Safety Information

MSDS https://www.manchesterorganics.com/R20498
Availability details for
Quantity
  • Availability
  • Add to Cart