MAN238931253 Manchester Organics

Imidazo[1,5-a]pyridine

  • MDL number MFCD03407375
  • Molecular Weight : 118.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P21255

Safety Information

MSDS https://www.manchesterorganics.com/P21255
Availability details for
Quantity
  • Availability
  • Add to Cart