MAN238931259 Manchester Organics

(R/S)-2-(3,4,5-Trimethoxyphenyl)butyric acid

  • MDL number MFCD06245369
  • Molecular Weight : 254.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B21007

Safety Information

MSDS https://www.manchesterorganics.com/B21007
Availability details for
Quantity
  • Availability
  • Add to Cart