MAN238931262 Manchester Organics

Ethyl 5-(2-methoxyphenyl)-5-oxopentanoate

  • MDL number MFCD01311632
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G21058

Safety Information

MSDS https://www.manchesterorganics.com/G21058
Availability details for
Quantity
  • Availability
  • Add to Cart