MAN238931270 Manchester Organics

2-(2-Pyridin-2-yl-1H-indol-3-yl)ethanamine monohydrochloride

  • MDL number MFCD06245347
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W21380

Safety Information

MSDS https://www.manchesterorganics.com/W21380
Availability details for
Quantity
  • Availability
  • Add to Cart