MAN238931293 Manchester Organics

1-Aminocycloheptane-1-carboxamide

  • MDL number MFCD10686671
  • Molecular Weight : 156.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L21133

Safety Information

MSDS https://www.manchesterorganics.com/L21133
Availability details for
Quantity
  • Availability
  • Add to Cart