MAN238931302 Manchester Organics

3-Chlorobenzene-1-carboximidamide hydrochloride

  • MDL number MFCD00276754
  • Molecular Weight : 191.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B21078

Safety Information

MSDS https://www.manchesterorganics.com/B21078
Availability details for
Quantity
  • Availability
  • Add to Cart