MAN238931351 Manchester Organics

3-(3-Hydroxyphenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11054000
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y20789

Safety Information

MSDS https://www.manchesterorganics.com/Y20789
Availability details for
Quantity
  • Availability
  • Add to Cart