MAN238931387 Manchester Organics

4-Diphenylaminoacetophenone

  • MDL number MFCD09695867
  • Molecular Weight : 287.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P21303

Safety Information

MSDS https://www.manchesterorganics.com/P21303
Availability details for
Quantity
  • Availability
  • Add to Cart