MAN238931389 Manchester Organics

4-Fluorobenzyl isothiocyanate

  • MDL number MFCD00041115
  • Molecular Weight : 167.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U21424

Safety Information

MSDS https://www.manchesterorganics.com/U21424
Availability details for
Quantity
  • Availability
  • Add to Cart