MAN238931401 Manchester Organics

3-(4-Hydroxyphenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11054003
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z20815

Safety Information

MSDS https://www.manchesterorganics.com/Z20815
Availability details for
Quantity
  • Availability
  • Add to Cart