MAN238931409 Manchester Organics

(3,4-Difluorophenyl)methanethiol

  • MDL number MFCD06798020
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A21194

Safety Information

MSDS https://www.manchesterorganics.com/A21194
Availability details for
Quantity
  • Availability
  • Add to Cart