MAN238931413 Manchester Organics

Ethyl ethoxyiminoacetate

  • MDL number MFCD06797654
  • Molecular Weight : 145.156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F21176
Availability details for
Quantity
  • Availability
  • Add to Cart