MAN238931449 Manchester Organics

3,6-Difluoro-2-methoxybenzaldehyde

  • MDL number MFCD08275289
  • Molecular Weight : 172.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G21319

Safety Information

MSDS https://www.manchesterorganics.com/G21319
Availability details for
Quantity
  • Availability
  • Add to Cart