MAN238931456 Manchester Organics

2,4-Difluoro-5-methylbenzonitrile

  • MDL number MFCD09835341
  • Molecular Weight : 153.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M21372

Safety Information

MSDS https://www.manchesterorganics.com/M21372
Availability details for
Quantity
  • Availability
  • Add to Cart