MAN238931457 Manchester Organics

4-Amino-6-chloropyrimidine-5-carbonitrile

  • MDL number MFCD07761812
  • Cas Number : 60025-09-4
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-6-chloro-5-pyrimidinecarbonitrile
Inchi Code 1S/C5H3ClN4/c6-4-3(1-7)5(8)10-2-9-4/h2H,(H2,8,9,10)
Inchi Key MAVMFCKRFRCMLE-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin GB
Product Link https://www.manchesterorganics.com/P21420
Availability details for
Quantity
  • Availability
  • Add to Cart