MAN238931501 Manchester Organics

5-Bromonicotinic acid ethyl ester

  • MDL number MFCD00040366
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/F21270
Availability details for
Quantity
  • Availability
  • Add to Cart