MAN238931502 Manchester Organics

4-Chloropyridine-3-sulfonamide

  • MDL number MFCD04038136
  • Molecular Weight : 192.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H21343

Safety Information

MSDS https://www.manchesterorganics.com/H21343
Availability details for
Quantity
  • Availability
  • Add to Cart