MAN238931516 Manchester Organics

3-Fluoro-4-methoxybenzylamine

  • MDL number MFCD04116360
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C21363

Safety Information

MSDS https://www.manchesterorganics.com/C21363
Availability details for
Quantity
  • Availability
  • Add to Cart