MAN238931517 Manchester Organics

2-Ethynyl-benzoic acid methyl ester

  • MDL number MFCD12547057
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/P20380
Availability details for
Quantity
  • Availability
  • Add to Cart