MAN238931518 Manchester Organics

4-Bromo-2-fluoropropoxybenzene

  • MDL number MFCD09749657
  • Molecular Weight : 233.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J21392

Safety Information

MSDS https://www.manchesterorganics.com/J21392
Availability details for
Quantity
  • Availability
  • Add to Cart