MAN238931526 Manchester Organics

2-Hydroxy-3-(trifluoromethyl)benzaldehyde

  • MDL number MFCD03788529
  • Molecular Weight : 190.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K21345
Availability details for
Quantity
  • Availability
  • Add to Cart