MAN238931528 Manchester Organics

2-Nitro-4-(trifluoromethyl)benzyl bromide

  • MDL number MFCD03095413
  • Molecular Weight : 284.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S21351
Availability details for
Quantity
  • Availability
  • Add to Cart