MAN238931530 Manchester Organics

4-Fluoro-2-(trifluoromethyl)phenol

  • MDL number MFCD00061290
  • Molecular Weight : 180.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y21808

Safety Information

MSDS https://www.manchesterorganics.com/Y21808
Availability details for
Quantity
  • Availability
  • Add to Cart