MAN238931578 Manchester Organics

2,4-Dichloro-5-nitro-6-methylpyrimidine

  • MDL number MFCD00023196
  • Molecular Weight : 208.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U21591

Safety Information

MSDS https://www.manchesterorganics.com/U21591
Availability details for
Quantity
  • Availability
  • Add to Cart