MAN238931582 Manchester Organics

2,4-Difluoro-5-methylbenzoic acid

  • MDL number MFCD09835285
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L21252
Availability details for
Quantity
  • Availability
  • Add to Cart