MAN238931600 Manchester Organics

2,6-Difluoro-4-iodophenol

  • MDL number MFCD09907951
  • Molecular Weight : 255.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M21443

Safety Information

MSDS https://www.manchesterorganics.com/M21443
Availability details for
Quantity
  • Availability
  • Add to Cart