MAN238931606 Manchester Organics

Indazole-7-boronic acid pinacol ester

  • MDL number MFCD08059264
  • Cas Number : 915411-02-8
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/H18858
Availability details for
Quantity
  • Availability
  • Add to Cart