MAN238931682 Manchester Organics

4-Chloro-2-fluorophenylacetic acid

  • MDL number MFCD03095402
  • Cas Number : 194240-75-0
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/H21320
Availability details for
Quantity
  • Availability
  • Add to Cart