MAN238931692 Manchester Organics

2-Hydroxy-3,5,6-trifluoropyridine

  • MDL number MFCD04114299
  • Molecular Weight : 149.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M19572

Safety Information

MSDS https://www.manchesterorganics.com/M19572
Availability details for
Quantity
  • Availability
  • Add to Cart