MAN238931699 Manchester Organics

4-Fluoro-2-nitrobenzonitrile

  • MDL number MFCD00277447
  • Molecular Weight : 166.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C21386

Safety Information

MSDS https://www.manchesterorganics.com/C21386
Availability details for
Quantity
  • Availability
  • Add to Cart