MAN238931707 Manchester Organics

3-Bromo-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD08059506
  • Molecular Weight : 253.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P21516

Safety Information

MSDS https://www.manchesterorganics.com/P21516
Availability details for
Quantity
  • Availability
  • Add to Cart