MAN238931708 Manchester Organics

2-Bromo-4,5-difluoroanisole

  • MDL number MFCD00070753
  • Molecular Weight : 223.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R21517

Safety Information

MSDS https://www.manchesterorganics.com/R21517
Availability details for
Quantity
  • Availability
  • Add to Cart