MAN238931713 Manchester Organics

2-Bromo-4,5-difluorotoluene

  • MDL number MFCD07777160
  • Cas Number : 875664-38-3
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z21358
Availability details for
Quantity
  • Availability
  • Add to Cart