MAN238931716 Manchester Organics

4-Bromo-2-(trifluoromethoxy)anisole

  • MDL number MFCD09907950
  • Cas Number : 853771-88-7
  • Molecular Weight : 271.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B21362
Availability details for
Quantity
  • Availability
  • Add to Cart