MAN238931755 Manchester Organics

4-Bromo-2-fluoro-5-(trifluoromethyl)aniline

  • MDL number MFCD04973757
  • Cas Number : 104460-70-0
  • Molecular Weight : 258.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T21494
Availability details for
Quantity
  • Availability
  • Add to Cart